missing translation for 'onlineSavingsMsg'
Learn More

Guar Gum, TRC

Product Code. 30240620
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240620 10 g 10g
30240720 5 g 5g
30240606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240620 Supplier Toronto Research Chemicals Supplier No. TRCG84471010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-30-0
Molecular Formula (C18H30O16)n
Synonym 1212A, AS 80, Activsoft S, Aglumix BR, BG 1, Burtonite V 7E, C 1000, C 1000 (gum), C 250, C 250 (gum), CG 70, CP 3300, CSA 200/50, CSA-M 175, CSAA-M 80, Celbond 7, Celca-Gum D 49D, Cyamopsis gum, DCA 9A, DX 9, Dabisco DHV 74, Dealca P 225, Dealca TP 1, Dealca TP 2, Decorpa, Duck Gum 800, Dycol 4500, E 412, EGMB, Ecopol 5060, Edicol ULV 50, Emcogum CSAA, Emulgum 200, Emulgum 200S, Galaxy 1083, Gendril Thik, Gendriv 162, Grindsted Guar 175, Guapack PF 20, Guapack PN, Guar, Guar 5200, Guar Fiber, Guar Gum FG 6070, Guar Gum GU/F, Guar Gum XS 5000, Guar HV 7000 CPS, Guar Pack PF 20, Guar Supercol U Fine, Guar WW250F, Guar flour, Guaran, Guarcel 302, Guarcol F 50, Guarcol U 40, Guargel D 15, Gum cyamopsis, Gum guar, Gums, guar, HR 71-51D, HS 30R, Herbapeck SF 08, Higum 551, Inagel GR 10, Inagel GR 10C, Indalca AG, Indalca AG-BV, Indalca AG-HV, J 2Fp, J 3000, J 3000 (polysaccharide), Jaguar, K 4492, KWL 2000, LGC 1, Lameprint DX 9, Lamgum 200, Lej Guar, Lipocard, Loloss, Lycoid DR, Meypro Guar CSAA M-100, Meypro Guar CSAA-M 175, Meypro-Guar 50, Meypro-Guar CSAA 200/50, Meypro-Guar CSAA-M 20, Meypro-Guar CSAA-M 225, Meyprodor 300, Meyprofin M 175, Meyprogat 30, Meyprogat 7, Meyprogum L, Meyprogum TC 47, Meyproid 6700, NGL 8158, Neosoft, Neovisco G, Newgelin G 100, Novatex SC 2, Oruno G 1, Oruno G 2, Papsize 7, Polycol WP 05, Polycol WP 10, Polycol WP 15, Polycol WP 20, Polytex 100, Procol F, Procol RF, Procol S 1, Procol U, RG 100, RG 1000, RT 3088, Rantec 4000, Rantec D 1, Rantec KP 4000, Regonol, Rein Guarin, SG, SG (guar gum), Sanbest NN 153, Seruparu SL 100, Soaguar RG 100, Stamulcol ULV 500, Super Tack, Supercol, Supergel 200, Supergel CSA, Supergel CSA 200/50, Supungeru GU 100, Syngum D 46D, TEGO Emulprot, Toshcol HV 77, Uni-Guar, Uniguar 80, Vidocrem A, Vidocreme A, Vidogum, Vofone WP, WG 15, WG 19, WG 1L, WGA 15, WGA 15L, WOGU 4401, X 5363, a-D-Galactopyrano-b-D-mannopyranan, a-D-galacto-b-D-Mannan

Specifications

Quantity 10 g
Chemical Name or Material Guar Gum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.