Learn More
Checkers Guard Dog™ Low Profile
Product Code. 17722418
Change view
Click to view available options
Length (Metric):
27 cm
43 cm
72 cm
77 cm
84 cm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
17722418 27 cm Each
17792408 43 cm Each
17732408 72 cm Each
17752408 77 cm Each
17782408 84 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17722418 Supplier Checkers Supplier No. PGDS0015OB
  • ONLINE EXCLUSIVE PRICE
    291.90€
    253.19€ / Pack of 1
    Save 38.71€
    13% Off
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market. These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
  • Guard Dog™ Low Profile models are the most compact top-loading cable protectors on the market
  • These protectors provide a safer crossing for vehicle and pedestrian traffic, while still maintaining a high load-bearing capacity for heavy equipment
Specifications
Length (English) 10.6 in.
Length (Metric) 27 cm
Width (Metric) 91 cm
Color Black, Orange
Material Polyurethane
Type Cable Protector