Learn More
Pacific Handy Cutter Guarded Spring Back Safety Cutter Left
Product Code. 17739037
Change view
Click to view available options
Color:
Green
Red
Unit Size:
Pack of 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17739037 Green Pack of 12
17729037 Red Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17739037 Supplier Pacific Handy Cutter Supplier No. S4SR
  • ONLINE EXCLUSIVE PRICE
    222.59€
    178.93€ / Pack of 12
    Save 43.66€
    19% Off
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The S4S is a safety knife with automatic blade retraction and additional protective guard. Therefore, it is very safe.
  • The protective guard also serves as a guide, e.g. when topping cartons.
  • With robust integrated tape splitter and spare blade storage.
  • Tool-free blade change.
  • With practical eyelet for lanyard fastening or hanging.
  • The green model is suitable for right-handed users.
  • The red model is suitable for left-handed users.
Specifications
Product Type Safety Cutter
Disposable Reusable
Sterility Non-sterile
Color Green