Learn More
Invitrogen™ GUCY1A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588718
Description
Immunogen sequence: MFCTKLKDLK ITGECPFSLL APGQVPNESS EEAAGSSESC KATVPICQDI PEKNIQESLP QRKTSRSRVY LHTLAESICK LIFPEFERLN VALQRTLAKH KIKESRKSLE REDFEKTIAE; Positive Samples: U-87MG, A-549, 293T, Mouse kidney, Mouse brain, Rat kidney, Rat spleen, Rat lung; Cellular Location: Cytoplasm.
Guanylate cyclases belong to the adenylyl cyclase class-4/guanylyl cyclase family. There are two forms of guanylate cyclase, a soluble form (GCS or sGC), which act as receptors for nitric oxide and a membrane-bound receptor form (GC), which are peptide hormone receptors. The GC-C protein is composed of an extracellular domain, a single transmembrane domain, and a cytoplasmic region consisting of a kinase-like domain and a catalytic domain. It is expressed as two differentially glycosylated forms, a 130 kDa precursor form present in the endoplasmic reticulum and a 145 kDa form present on the plasma membrane. Ligand binding to the extracellular domain of GC-C promotes the accumulation of cGMP. GC-C acts as the receptor for heatstable enterotoxins, small peptides secreted by some pathogenic strains of E. coli that cause severe secretory diarrhea. GC-C also binds to guanylin and uroguanylin peptides, which modulate renal function in response to oral salt load.
Specifications
| GUCY1A3 | |
| Polyclonal | |
| Unconjugated | |
| GUCY1A1 | |
| 1200016O07Rik; 4.6.1.2; alpha 1 sGC; GC-SA3; GCSalpha1; GC-S-alpha-1; GCS-alpha1; GCS-alpha-1; GCS-alpha-3; guanylate cyclase 1 soluble subunit alpha; guanylate cyclase 1, soluble, alpha 3; Guanylate cyclase soluble alpha 1 (GTP pyrophosphate - lyase); Guanylate cyclase soluble subunit alpha-1; Guanylate cyclase soluble subunit alpha-3; Guanylate cyclase, soluble, alpha 1 (GTP pyrophosphate - lyase); GUC1A3; GUCA3; GUCSA3; Gucy1a1; GUCY1A3; MYMY6; SGC; sGC-alpha1; soluble guanylate cyclase large subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2982, 497757, 60596 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P19686, Q02108, Q9ERL9 | |
| GUCY1A1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human GUCY1A3 (NP_0008472). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.