missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GW 501516
GREENER_CHOICE

Product Code. 18753168
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18753168 10 mg 10mg
18794719 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18753168 Supplier Tocris Bioscience™ Supplier No. 5674/10
 Restricted Item
Estimated Shipment: 23-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective PPARδ agonist

GW 501516 is a potent and selective PPARδ agonist (EC50 = 1.2 nM). Displays <1000-fold selectivity over other PPAR subtypes. Increases ABC A1 transporter expression and induces apolipoprotein A1-mediated cholesterol efflux in vitro. Also increase serum HDL cholesterol and lowers small, dense LDL levels in obesity in vivo models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]acetic acid
CAS 317318-70-0
Quantity 10 mg
Target PPAR delta Receptor Agonists
Molecular Formula C21H18F3NO3S2
Formula Weight 453.5
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.