missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ H-2Dd Monoclonal Antibody (34-5-8S), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ A15444
This item is not returnable.
View return policy
Description
H-2Dd Monoclonal Antibody for Flow
Involved in the presentation of foreign antigens to the immune system. [UniProt] HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| H-2Dd | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.4 | |
| P01895, P01896, P01897, P01899, P01900, P14426, P14427 | |
| H2-D1 | |
| mouse H-2Dd from BDF1 mouse splenocytes. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry | |
| 34-5-8S | |
| FITC | |
| H2-D1 | |
| H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2-DC1-beta transplantation antigen protein; H2DD; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein | |
| Mouse | |
| purified | |
| RUO | |
| 14964 | |
| 4°C, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction