missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Kq Mouse anti-Mouse, BV750, Clone: KH114, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747104
This item is not returnable.
View return policy
Description
H-2Kq Monoclonal antibody specifically detects H-2Kq in Mouse samples. It is validated for Flow Cytometry
Specifications
| H-2Kq | |
| Monoclonal | |
| Brilliant Violet 750 | |
| H-2Kq; H2-Kq; Histocompatibility-2Kq | |
| B10.Q mouse skin graft and splenocytes | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| KH114 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction