Learn More
H-Ala-Ala-Ala-Ala-Ala-OH, TRC
Product Code. 30326303
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326303 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326303 Supplier Toronto Research Chemicals Supplier No. TRCA5761202.5MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material H-Ala-Ala-Ala-Ala-Ala-OH
CAS 10183-34-3
Molecular Formula C15H27N5O6
Quantity 2.5 mg
Synonym Penta-L-alanine, AAAAA (peptide), Alanine, N-[N-[N-(N-L-alanyl-L-alanyl)-L-alanyl]-L-alanyl]- (6CI,7CI), Alanine, N-[N-[N-(N-L-alanyl-L-alanyl)-L-alanyl]-L-alanyl]-, L- (8CI), L-Alanine, N-[N-[N-(N-L-alanyl-L-alanyl)-L-alanyl]-L-alanyl]-, L-Alanyl-L-alanyl-L-alanyl-L-alanyl-L-alanine
InChI Formula InChI=1S/C15H27N5O6/c1-6(16)11(21)17-7(2)12(22)18-8(3)13(23)19-9(4)14(24)20-10(5)15(25)26/h6-10H,16H2,1-5H3,(H,17,21)(H,18,22)(H,19,23)(H,20,24)(H,25,26)/t6-,7-,8-,9-,10-/m0/s1
SMILES C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)O
IUPAC Name (2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-aminopropanoyl]amino]propanoyl]amino]propanoyl]amino]propanoyl]amino]propanoic acid
Molecular Weight (g/mol) 373.4
Formula Weight 373.1961