Learn More
Thermo Scientific™ H-Flex HS4 Swinging Bucket Rotor with SlideCoat for 4 L General Purpose X Pro and ST Plus Centrifuges
Shop All Thermo Scientific ProductsDescription
Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor at 7197xg in 4 L High Performance General Purpose Centrifuges .
- Auto-Lock enable fast, one-handed rotor exchanges
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Specifications
| Material | Anodized Aluminum |
| Type | Swinging Bucket Rotor |
| Capacity | 16 x 50 mL or 36 x 15 mL, 66 x 10 mL or 84 x 5/7 mL Blood Tubes |
| Max. RCF | 7,197 x g |
| Max. Speed | 6,100 rpm |
| For Use With (Equipment) | Sorvall X4 Pro, Sorvall X4 F Pro, Multifuge X4 Pro, Multifuge X4 F Pro, Sorvall ST4 Plus, Sorvall ST4F Plus, Megafuge ST4 Plus, Megafuge ST4F Plus |
| Includes | Windshield, rotor cross and buckets, 10 microwell microplates, 2 deepwell plates |
| Biocontainment | Yes |
| Autoclavable | Autoclavable |