missing translation for 'onlineSavingsMsg'
Learn More

H-Glu(OBzl)-OH, TRC

Product Code. 30390280
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390280 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390280 Supplier Toronto Research Chemicals Supplier No. TRCG598170500MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material H-Glu(OBzl)-OH
CAS 1676-73-9
Molecular Formula C42 H70 O11
Quantity 500 mg
Synonym Benzyl glutamate, (2S)-2-Amino-5-(benzyloxy)-5-oxopentanoic acid, (S)-2-Amino-5-(benzyloxy)-5-oxopentanoic acid, 5-Benzyl L-glutamate, gamma-Benzyl L-glutamate, gamma-Benzyl glutamate, gamma-Benzylglutamic acid, NSC 9969
InChI Formula InChI=1S/C42H70O11/c1-11-29(38(46)47)31-15-14-23(4)36(50-31)27(8)34(44)26(7)35(45)30(12-2)37-24(5)22-25(6)41(51-37)19-16-32(43)42(53-41)21-20-39(10,52-42)33-17-18-40(48,13-3)28(9)49-33/h16,19,23-34,36-37,43-44,48H,11-15,17-18,20-22H2,1-10H3,(H,46,47)/t23-,24-,25+,26-,27+,28+,29+,30-,31+,32-,33-,34-,36-,37-,39+,40?,41+,42?/m1/s1
SMILES CC[C@@H]([C@@H]1CC[C@@H](C)[C@@H](O1)[C@@H](C)[C@H](O)[C@@H](C)C(=O)[C@@H](CC)[C@@H]2O[C@@]3(OC4(CC[C@](C)(O4)[C@H]5CCC(O)(CC)[C@H](C)O5)[C@H](O)C=C3)[C@@H](C)C[C@H]2C)C(=O)O
IUPAC Name (2S)-2-[(2S,5R,6R)-6-[(2S,3S,4R,6S)-6-[(3S,7R,9R,10R,12S,15R)-3-[(2R,6S)-5-ethyl-5-hydroxy-6-methyloxan-2-yl]-15-hydroxy-3,10,12-trimethyl-4,6,8-trioxadispiro[4.1.57.35]pentadec-13-en-9-yl]-3-hydroxy-4-methyl-5-oxooctan-2-yl]-5-methyloxan-2-yl]butanoic acid
Molecular Weight (g/mol) 750.999
Formula Weight 750.492
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.