missing translation for 'onlineSavingsMsg'
Learn More

H-Gly-Tyr-OH, 98%, BLD Pharm™

Product Code. 31423896
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31423896 5 g
31421986 100 g
31416806 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31423896 Supplier BLD Pharm Supplier No. BD132495g
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 658-79-7
Molecular Formula C11H14N2O4
MDL Number MFCD00008126
Quantity 5 g
SMILES O=C(N[C@@H](CC1=CC=C(C=C1)O)C(O)=O)CN
Molecular Weight (g/mol) 238.24
Percent Purity 98%
Chemical Name or Material H-Gly-Tyr-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.