missing translation for 'onlineSavingsMsg'
Learn More

H-Val-Met-OH, TRC

Product Code. 30451010
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451010 50 mg -
30457278 100 mg -
30457311 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30451010 Supplier Toronto Research Chemicals Supplier No. TRCV99170550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14486-09-0
Molecular Formula C10H20N2O3S
Molecular Weight (g/mol) 248.34
IUPAC Name (2S)-2-[[(2S)-2-amino-3-methylbutanoyl]amino]-4-methylsulfanylbutanoic acid
SMILES CSCC[C@H](NC(=O)[C@@H](N)C(C)C)C(=O)O

Specifications

InChI Formula InChI=1S/C10H20N2O3S/c1-6(2)8(11)9(13)12-7(10(14)15)4-5-16-3/h6-8H,4-5,11H2,1-3H3,(H,12,13)(H,14,15)/t7-,8-/m0/s1
Quantity 50 mg
Formula Weight 248.1195
Shipping Condition Room Temperature
Chemical Name or Material H-Val-Met-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.