Learn More
R&D Systems™ H3N2 Neuraminidase (AAO46464.1) VersaClone cDNA
Specifications
Specifications
| Gene | Influenza A H3N2 Neuraminidase |
| Accession Number | AAO46464.1 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |