missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H60 Antibody (205326) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1155JF635
This item is not returnable.
View return policy
Description
H60 Monoclonal antibody specifically detects H60 in Mouse samples. It is validated for Flow Cytometry,Western Blot,Blockade of Receptor-ligand Interaction,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Specifications
| H60 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| H60a | |
| BaF3 mouse pro-B cell line transfected with mouse H60 and mouse myeloma cell line NS0-derived recombinant mouse H60, Asp30-Gln212, Accession # Q3TDZ7 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Western Blot, Blockade of Receptor-ligand Interaction, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 205326 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.5 mL | |
| Cancer | |
| 15101 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction