missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HADHA Monoclonal Antibody (24GB6655)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA557420

 View more versions of this product

Product Code. 30275513

  • 564.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HADHA Monoclonal Antibody for Western Blot

This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HADHA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P40939, Q64428, Q8BMS1
HADHA
Recombinant protein of human HADHA (aa 106-325).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Western Blot
24GB6655
Unconjugated
HADHA
3-ketoacyl-Coenzyme A (CoA) thiolase, alpha subunit; 3-oxoacyl-CoA thiolase; 78 kDa gastrin-binding protein; C77020; ECHA; gastrin-binding protein; GBP; HADH; HADHA; hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit; LCEH; LCHAD; Long chain 3-hydroxyacyl-CoA dehydrogenase; long-chain 2-enoyl-CoA hydratase; Long-chain enoyl-CoA hydratase; long-chain-3-hydroxyacyl-CoA dehydrogenase; mitochondrial long-chain 2-enoyl-Coenzyme A (CoA) hydratase, alpha subunit; mitochondrial long-chain L-3-hydroxyacyl-Coenzyme A (CoA) dehydrogenase, alpha subunit; mitochondrial trifunctional enzyme, alpha subunit; mitochondrial trifunctional protein, alpha subunit; Monolysocardiolipin acyltransferase; Mtpa; RGD1560655; TP-alpha; Trifunctional enzyme subunit alpha, mitochondrial
Mouse
Affinity chromatography
RUO
170670, 3030, 97212
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.