missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-1/HGFA Inhibitor 1 Antibody (028), FITC , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89974F
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Monoclonal antibody specifically detects HAI-1/HGFA Inhibitor 1 in Human samples. It is validated for ELISA
Specifications
| HAI-1/HGFA Inhibitor 1 | |
| Monoclonal | |
| FITC | |
| HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HAI-1/HGFA Inhibitor 1 (O43278-2; Met1-Val433). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 028 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 6692 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction