Learn More
HAI-1 Mouse anti-Human, Unconjugated, Clone: 2H2D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67031-1-IG-150UL
Description
The HAI-1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HAI-1. This antibody recognizes human antigen. The HAI-1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| HAI-1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 2H2D8 | |
| Unconjugated | |
| 150 μL | |
| 6692 | |
| IgG1 |