Learn More
Haloacetic acid mixture for HJ 758-2015 various concentrations in Methyl tert-butyl ether, Dr. Ehrenstorfer
Product Code. 30311033
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30311033 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30311033 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000548MB
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 79-08-3, 5589-96-8, 71133-14-7, 79-11-8, 75-99-0, 631-64-1
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Product Title
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.