missing translation for 'onlineSavingsMsg'
Learn More

Halofuginone Hydrochloride, TRC

Product Code. 18062830
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062830 5 mg 5mg
18050920 10 mg 10mg
18003830 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18062830 Supplier Toronto Research Chemicals Supplier No. TRCH1025005MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1217623-74-9
Molecular Formula C16 H17 Br Cl N3 O3 . Cl H
Molecular Weight (g/mol) 451.14
IUPAC Name 7-bromo-6-chloro-3-[3-[(2S,3R)-3-hydroxypiperidin-2-yl]-2-oxopropyl]quinazolin-4-one;hydrochloride
SMILES Cl.O[C@@H]1CCCN[C@H]1CC(=O)CN2C=Nc3cc(Br)c(Cl)cc3C2=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H17BrClN3O3.ClH/c17-11-6-13-10(5-12(11)18)16(24)21(8-20-13)7-9(22)4-14-15(23)2-1-3-19-14;/h5-6,8,14-15,19,23H,1-4,7H2;1H/t14-,15+;/m0./s1
Formula Weight 448.9909
Percent Purity >95%
Chemical Name or Material Halofuginone Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.