Learn More
Olympus™ Halogen Lamp
Product Code. 10436382
Change view
Click to view available options
Wattage:
30 W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wattage unitSize
10436382 30 W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10436382 Supplier Olympus™ Supplier No. E0430150
  • ONLINE EXCLUSIVE PRICE
    39.15€
    10.24€ / Pack of 1
    Save 28.91€
    73% Off
Estimated Shipment: 03-11-2026
Specifications
Voltage 6 V
Wattage 30 W
For Use With (Equipment) CX31 / CX41 / BX41ECO ET BX41ERGO
Product Type Halogen Lamp