Learn More
Hamilton™ Chemically Inert Manual Valves
Product Code. 10217362
Change view
Click to view available options
Type:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10217362 3-2 Manual Valve Each
10337272 3-3 Manual Valve Each
11562425 1-1 Manual Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10217362 Supplier Hamilton™ Supplier No. 86728
  • ONLINE EXCLUSIVE PRICE
    141.00€
    126.48€ / Pack of 1
    Save 14.52€
    10% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Valves have a fluid path containing PTFE and CTFE
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifications
Material CTFE, PTFE
Type 3-2 Manual Valve