Learn More
Hamilton™ Kel-F Hub Sharp Non-Coring Needles (Luer Lock)
Product Code. 11592275
Change view
Click to view available options
Needle Gauge:
18 G
20 G
21 G
22 G
22s G
23 G
25 G
26 G
26s G
Needle Point Style:
Sharp, Bevel, Curved
Tapered
Diameter (English) Inner:
0.005 in.
0.01 in.
0.013 in.
0.016 in.
0.02 in.
0.033 in.
Diameter (English) Outer:
0.01 in.
0.018 in.
0.025 in.
0.028 in.
0.032 in.
0.035 in.
0.05 in.
Förpackningsstorlek:
Each
Pack of 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Needle Gauge Needle Point Style Diameter (English) Inner Diameter (English) Outer unitSize
11592275 21 G Sharp, Bevel, Curved 0.02 in. 0.032 in. Pack of 6
11582275 20 G Sharp, Bevel, Curved 0.02 in. 0.035 in. Pack of 6
11562275 18 G Sharp, Bevel, Curved 0.033 in. 0.05 in. Pack of 6
15186764 23 G Sharp, Bevel, Curved 0.013 in. 0.025 in. Each
11502285 23 G Sharp, Bevel, Curved 0.013 in. 0.025 in. Pack of 6
11522285 25 G Sharp, Bevel, Curved 0.01 in. 0.01 in. Pack of 6
11532285 26 G Sharp, Bevel, Curved 0.01 in. 0.018 in. Pack of 6
11582285 26s G Tapered 0.005 in. 0.018 in. Pack of 6
10008340 22s G Tapered 0.005 in. 0.028 in. Pack of 6
10363421 22 G Sharp, Bevel, Curved 0.016 in. 0.028 in. Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11592275 Leverantör Hamilton™ Leverantörsnummer 90121
Bara null kvar
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
Kel-F hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2 in. (51 mm). Custom lengths and point styles are available.
Kel-F Hub Compatible Syringe Terminations
  • Luer Tip (LT)- The needles are removable and fit over a ground glass hub which is tapered in the shape of a male luer. The LT termination will accept most hypodermic needles but was designed specifically for use with Hamilton Kel-F needles. This termination increases the dead volume in the syringe, which may not be appropriate for some applications. Autoclavable when disassembled.
  • PTFE Luer Lock (TLL)- This termination has a PTFE, male Luer taper with nickelplated brass locking hub for use with Kel-F needles, metal hub needles, and universal connectors. Also, the TLL is used with Hamilton diluters/dispensers, OEM applications, and manual operations. The version with slots is used during the attachment to some instruments but does not effect the connection to needles. Autoclavable when disassembled, except on 25 mL and greater syringes. Repeated autoclaving will shorten syringe life.
Needle Point Styles
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 31-10
Six needles per pack.
Specifikationer
Autoclavable Not Autoclavable
Material Stainless Steel
Needle Gauge 21 G
Diameter (English) Inner 0.02 in.
Diameter (English) Outer 0.032 in.
Diameter (Metric) Inner 0.51 mm
Diameter (Metric) Outer 0.82 mm
Length (English) Needle 2 in.
Product Type Kel-F Hub Needle
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved
For Use With (Equipment) Luer-tip and PTFE Luer-lock Syringes and Connectors
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.