Learn More
Thermo Scientific™ Hand Dispenser Kit
Product Code. 15102893 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Hand Dispenser Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
15102893 Hand Dispenser Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15102893 Supplier Thermo Scientific™ Supplier No. 50138221
  • ONLINE EXCLUSIVE PRICE
    1195.00€
    1117.00€ / Pack of 1
    Save 78.00€
    6% Off
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Increase lab flexibility with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems.
Increase flexibility in the lab with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems. This kit includes a dispenser with 3 meter cord that connects to the water system tank and a 0.2 micron final filter. It is compatible with Barnstead LabTower™, Pacific™ and Smart2Pure™ systems.
Optional for the following Barnstead water purification systems:
  • LabTower™ EDI
  • LabTower™ RO
  • LabTower™ TII
  • Smart2Pure™
  • Pacific™ TII + tank (tank must have optional pump)
  • Pacific™ RO + tank (tank must have optional pump)
TRUSTED_SUSTAINABILITY
Specifications
Description Hand Dispenser Kit
For Use With (Equipment) LabTower EDI, LabTower TII, Smart2Pure, Pacific TII + Tank (Tank Must Have Optional Pump), Pacific RO + Tank (Tank Must Have Optional Pump), LabTower RO
Includes Hand Dispenser Kit (includes hand dispenser with 3 meter cord, and 0.2 micron final filter)