missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAND1 Antibody (8E7A11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
187.00€
Specifications
| Antigen | HAND1 |
|---|---|
| Clone | 8E7A11 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
HAND1 Monoclonal specifically detects HAND1 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
| HAND1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| BHLHA27, bHLHa27heart- and neural crest derivatives-expressed protein 1, Class A basic helix-loop-helix protein 27, EHAND, Extraembryonic tissues, heart, autonomic nervous system and neural crestderivatives-expressed protein 1, heart and neural crest derivatives expressed 1, Hxt, Thing1 | |
| HAND1 | |
| IgG1 | |
| Unpurified |
| 8E7A11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 9421 | |
| Purified recombinant fragment of HAND1 (aa90-190) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title