Learn More
Brady™ HandiMark™ Indoor/Outdoor Vinyl Tape
Description
- Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
- The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
- Handimark Consumables Equivalence for BMP71 printers: M71C-500-595-WT (see Related Items).
- Handimark Consumables Equivalence for BMP61 and M611 printers: M61C-500-595-WT (see Related Items).
Specifications
Specifications
| Certifications/Compliance | RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC |
| Description | HandiMark Indoor/Outdoor Vinyl Tape |
| Adhesive Type | Permanent Acrylic, Permanent |
| Coating | Powder |
| Color | Red |
| Finish | Gloss |
| Material | Vinyl |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| For Use With (Application) | Component and equipment labelling, Electrical labelling, Facility Identification, Floor marking, Laboratory labelling, Lean and 5S labelling, Pipe and valve marking, Warehouse marking |
| Show More |