Learn More
DuraLab Hanging Bucket with Clip-On Lid for Laboratory Trolleys
Product Code. 30280259
Change view
Click to view available options
Product Type:
Basket
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
30280259 Basket Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30280259 Supplier DuraLab Supplier No. DRL0013
  • ONLINE EXCLUSIVE PRICE
    22.05€
    20.45€ / Pack of 1
    Save 1.60€
    7% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A bucket with a clip-on lid designed to work with the DuraLab Laboratory Trolley range.
  • Attaches to the ends of DuraLab Laboratory Trolleys for convenient placement
  • Clip-on lid keeps contents contained while allowing easy removal
  • Made from durable Polyethylene (PE) material for reliable use
  • Can be used for storing items or collecting waste in laboratory settings
  • Lightweight design allows easy handling and movement
Specifications
Product Type Basket
Color Black
Not compatible with DuraLab Laboratory Trolleys with deep shelves.