missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Haptoglobin Antibody (KHK patent anti-Haptoglobin) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28166-1mg
This item is not returnable.
View return policy
Description
Haptoglobin Monoclonal antibody specifically detects Haptoglobin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Haptoglobin | |
| Monoclonal | |
| Unconjugated | |
| P00738 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| KHK patent anti-Haptoglobin | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| binding peptide, BP, haptoglobin, haptoglobin alpha(1S)-beta, haptoglobin alpha(2FS)-beta, haptoglobin, alpha polypeptide, haptoglobin, beta polypeptide, HP2ALPHA2, HPA1S, MGC111141, zonulin | |
| Haptoglobin | |
| 1 mg | |
| Cardiovascular Biology | |
| 3240 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction