missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Haptoglobin Monoclonal Antibody (C6), Invitrogen™

Mouse Monoclonal Antibody
134.00€ - 453.00€
Specifications
| Antigen | Haptoglobin |
|---|---|
| Clone | C12 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17694236
|
Invitrogen™
MA543785 |
20 μL |
134.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17684236
![]() |
Invitrogen™
MA543784 |
100 μL |
453.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Haptoglobin is an a2-globulin plasma protein synthesized in the liver. It is an acute phase protein which also acts as a scavenger of hemoglobin from hemolyzed red cells. The haptoglobin monomer consists of two a and two beta chains; the a chains bay be hp1Fa, hp1Sa, or hp2a types while the beta chain is constant. Three major phenotypes are recognized: HP1-1, Hp2-1 and Hp-2-2, in which the hp1a chain bay be either the F or S type.Specifications
| Haptoglobin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| alpha polypeptide; Ba1-647; beta polypeptide; binding peptide; Binding peptide (BP); BP; haptoglobin; haptoglobin alpha 1S; Haptoglobin alpha chain; Haptoglobin alpha chain / alpha(1S) beta; haptoglobin alpha(1S)-beta; Haptoglobin alpha(2FS) beta; haptoglobin alpha(2FS)-beta; Haptoglobin beta chain; haptoglobin, alpha polypeptide; haptoglobin, beta polypeptide; haptoglobin; haptoglobin-like; haptoglobin-like; haptoglobin-related protein; Hp; HP-1; HP2 ALPHA2; HP2ALPHA2; HP2-ALPHA-2; HPA1S; HPR; HPT; Liver regeneration-related protein LRRG173; LOC100067869; LOC479668; MGC111141; MGC133752; preHP2; zonulin | |
| HP | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C12 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P00738, P06866 | |
| 24464, 3240 | |
| Recombinant Hpt (Lys227-Asp378) expressed in E.coli. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| HP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title