Learn More
Horst™ HB Model Heating Tape
Product Code. 10301992
Change view
Click to view available options
Length (Metric):
0.5 m
2.5 m
3 m
4 m
5 m
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10301992 4 m Each
10637892 5 m Each
10210592 0.5 m Each
10341892 2.5 m Each
10427511 3 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10301992 Supplier Horst™ Supplier No. 20211
  • ONLINE EXCLUSIVE PRICE
    300.30€
    199.31€ / Pack of 1
    Save 100.99€
    33% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Dimensions (L x W) 4 Lm
Dimensions (L x W x H) 4 Lm
Length (Metric) 4 m
Voltage 230 V
Electrical Requirements ∼230V Voltage
Model HB
Wattage 900w
Width (Metric) 30mm
Product Type Heating Tape
Temperature (Metric) 450°C