Learn More
Horst™ HB Model Heating Tape
Product Code. 10637892
Change view
Click to view available options
Length (Metric):
0.5 m
2.5 m
3 m
4 m
5 m
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10637892 5 m Each
10210592 0.5 m Each
10301992 4 m Each
10341892 2.5 m Each
10427511 3 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10637892 Supplier Horst™ Supplier No. 20207
  • ONLINE EXCLUSIVE PRICE
    396.00€
    236.43€ / Pack of 1
    Save 159.57€
    40% Off
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Dimensions (L x W) 5 Lm
Dimensions (L x W x H) 5 Lm
Length (Metric) 5 m
Electrical Requirements ∼230V Voltage
Model HB
Product Type Heating Tape