missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HBB Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543781
This item is not returnable.
View return policy
Description
The alpha (HBA) and beta (HBB) loci determine the structure of the 2 types of polypeptide chains in adult hemoglobin, Hb A. The normal adult hemoglobin tetramer consists of two alpha chains and two beta chains. Mutant beta globin causes sickle cell anemia. Absence of beta chain causes beta-zero-thalassemia. Reduced amounts of detectable beta globin causes beta-plus-thalassemia. The order of the genes in the beta-globin cluster is 5'-epsilon-gamma-G-gamma-A-delta-beta-3'.
Specifications
| HBB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P68871 | |
| HBB | |
| Met1-His147 (Accession P68871), with N-terminal His and GST Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| C3 | |
| Unconjugated | |
| HBB | |
| beta globin chain; beta-1-globin; Beta-globin; CD113t-C; Hbb; hemoglobin beta; Hemoglobin beta chain; hemoglobin beta chain complex; hemoglobin beta chain, major-form; Hemoglobin beta-1 chain; hemoglobin subunit beta; Hemoglobin subunit beta-1; hemoglobin, beta; III beta-1 globin; III beta-2 globin; LVV-hemorphin-7; Spinorphin | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3043 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction