missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ HBM™ Basal Medium, 500 mL

Product Code. 11695450
Click to view available options
Quantity:
500 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11695450

Brand: Lonza™ CC3199

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Basal medium for use with HCM™ SingleQuots™ (CC-4182) growth supplement pack to support primary hepatocytes from all species.

  • Basal medium.
  • Contains no growth factors cytokines, or supplements.
  • For use with HCM™ SingleQuots™ (CC-4182) growth supplement pack.
  • In combination with supplements, this medium make a highly reproducible maintenance medium for primary hepatocytes from all species.
  • Final medium is serum free.
  • Optimized for hepatocyte cell health and compatible with all species hepatocytes.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hepatocyte Cells
Product Type Basal Medium
Quantity 500 mL
pH 7.2 to 7.6
Color Clear
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.