Learn More
Zeiss™ HBO 100 Illuminator
Product Code. 11835440
Change view
Click to view available options
Type:
HBO 100 Illuminator
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
11835440 HBO 100 Illuminator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11835440 Supplier Zeiss™ Supplier No. 4230119901000
  • ONLINE EXCLUSIVE PRICE
    5890.00€
    3883.34€ / Pack of 1
    Save 2006.66€
    34% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Features automatic lamp alignment
  • HBO 100 illuminator
  • With collector and lamp mount
  • Includes heat-reflecting filter
  • Automatic Lamp Alignment
Specifications
Type HBO 100 Illuminator
Description Features automatic lamp alignment
For Use With (Equipment) Zeiss Microscopes
Includes Lamp Mount and Collector