Learn More
Horst™ HBQ Model Very Flexible Heating Tape
Product Code. 10148781
Change view
Click to view available options
Length (Metric):
0.5 m
1 m
1.5 m
2 m
3 m
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10148781 3 m Each
10332322 0.5 m Each
10087141 1 m Each
10322462 1.5 m Each
10486371 2 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10148781 Supplier Horst™ Supplier No. 20406
  • ONLINE EXCLUSIVE PRICE
    889.01€
    554.36€ / Pack of 1
    Save 334.65€
    37% Off
View all promotions
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 3 Lm
Dimensions (L x W x H) 3 Lm
Length (Metric) 3 m
Electrical Requirements ∼230V Voltage
Model HBQ
Product Type Very Flexible Heating Tape