Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red
GREENER_CHOICE
Product Code. 11530476 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
10 x 500 mL
20 x 100 mL
Unit Size:
100mL
2000mL
5000mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11530476 500 mL 500mL
11520476 100 mL 100mL
11550476 20 x 100 mL 2000mL
11540476 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11530476 Supplier Gibco™ Supplier No. 14175053
 Restricted Item
In Stock
Add to basket
Add to basket
HBSS, no calcium, no magnesium, no phenol red
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
cGMP Manufacturing and Quality System
Gibco™ HBSS is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ HBSS product made in our Grand Island facility. This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 15 to 30°C
Description Hanks' Balanced Salt Solution, 500mL, Liquid, 1 X Concentration, 6.7 to 7.8 pH, With Glucose, Sodium Bicarbonate, 36 Months Shelf Life, For Mammalian Cell Culture
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
With Additives Glucose, Sodium Bicarbonate
Without Additives No Calcium, No Magnesium, No Phenol Red
For Use With (Application) Mammalian Cell Culture
pH 6.7 to 7.8
Quantity 500 mL
Regulatory Status In-vitro Diagnostic Use