Learn More
Horst™ HBST Model Heating Tape
Product Code. 10501742
Change view
Click to view available options
Length (Metric):
1 m
2 m
3 m
4 m
5 m
10 m
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10501742 10 m Each
10057071 1 m Each
10699802 2 m Each
10590392 3 m Each
10077081 4 m Each
10751753 5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10501742 Supplier Horst™ Supplier No. 20706
  • ONLINE EXCLUSIVE PRICE
    500.98€
    312.21€ / Pack of 1
    Save 188.77€
    37% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Dimensions (L x W) 10 Lm
Dimensions (L x W x H) 10 Lm
Length (Metric) 10 m
Electrical Requirements ∼230V Voltage
Model HBST
Product Type Heating Tape