Learn More
Taylor Wharton™ HC35 High Capacity Cryostorage System
Product Code. 11728519
Change view
Click to view available options
Capacity (Metric):
35 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11728519 35 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11728519 Supplier Taylor Wharton™ Supplier No. HC35
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 31-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ruggedly constructed, high-capacity cryogenic storage dewar. Taylor Wharton™ HC35 High Capacity Cryostorage System uses a convenient canister index location ring and a tapered internal spider.
  • Large storage capacity.
  • New Rugged Construction - ribbed high strength aluminum body, magniformed necktube design, and more durable paint.
  • Versatile Storage System - convenient canister index location ring and tapered internal spider.
  • Superior vacuum performance with super insulation provides maximum holding times.
  • Security - Optional low-level alarm is available with remote monitor capability.
  • Mobility - Roller bases are available for all models
Specifications
Height (Metric) Exterior 681 mm
Includes 10 canisters, 67 x 279mm
Weight (Metric) 17.7/46 kg (empty/full)
Width (Metric) Exterior 478 mm
Capacity (Metric) 35 L
Holds Vials (1020 x 2 mL)