missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCLS1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04028UV
This item is not returnable.
View return policy
Description
HCLS1 Polyclonal antibody specifically detects HCLS1 in Human samples. It is validated for Western Blot
Specifications
| HCLS1 | |
| Polyclonal | |
| Western Blot | |
| CTTNL, hematopoietic cell-specific Lyn substrate 1hematopoietic lineage cell-specific protein, HS1lckBP1, LckBP1, p75 | |
| The immunogen recognized by this antibody maps to a region between residue 350 and 400 of human Hematopoietic Cell-specific Lyn Substrate 1 using the numbering given in entry NP_005326.2 (GeneID 3059). | |
| 0.1 mL | |
| Signal Transduction | |
| 3059 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction