missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC1 Antibody (CL0510) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52937JF669
This item is not returnable.
View return policy
Description
HDAC1 Monoclonal antibody specifically detects HDAC1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| HDAC1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 3.5.1.98, GON-10, HD1DKFZp686H12203, histone deacetylase 1, reduced potassium dependency, yeast homolog-like 1, RPD3L1RPD3 | |
| This antibody was developed using a recombinant protein derived from Q13547, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 3065 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0510 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction