Learn More
Invitrogen™ HDAC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1860
Description
PA1-860 detects histone deacetylase 1 (HDAC1) from human, mouse, rat, hamster and canine tissues and cells. PA1-860 has been successfully used in Western blot, immunoprecipitation, ChIP, immunofluoresence, immunocytochemistry and immunohistochemistry procedures. By Western blot, this antibody detects an ~62 kDa protein representing HDAC1 from HeLa cell lysate. Immunocytochemical staining of HDAC1 in HeLa cells with PA1-860 results in nuclear staining. The PA1-860 immunogen is a synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1. This immunizing peptide (Cat. # PEP-069) is available for use in neutralization and control experiments.
Histone acetylation and deacetylation, catalyzed by multisubunit complexes, play a key role in the regulation of eukaryotic gene expression. HDAC1 belongs to the histone deacetylase/acuc/apha family and is a component of the histone deacetylase complex. It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation. Together with metastasis-associated protein-2, HDAC1 deacetylates p53 and modulates its effect on cell growth and apoptosis.
Specifications
| HDAC1 | |
| Polyclonal | |
| Unconjugated | |
| HDAC1 | |
| DKFZp686H12203; GON-10; HD1; Hdac1; Hdac1-ps; Histone deacetylase 1; histone deacetylase 1-like protein; MommeD5; reduced potassium dependency, yeast homolog-like 1; RP4-811H24.2; RPD3; RPD3L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100752721, 297893, 3065, 433759, 487309 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O09106, Q13547, Q4QQW4 | |
| HDAC1 | |
| Synthetic peptide corresponding to residues C E(467) E K P E A K G V K E E V K L A(482) of human HDAC1. | |
| 100 μg | |
| Primary | |
| Canine, Hamster, Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.