Learn More
HDAC2 Mouse anti-Human, Unconjugated, Clone: 1A3E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67165-1-IG-150UL
Description
The HDAC2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HDAC2. This antibody recognizes human, mouse, rat antigen. The HDAC2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| HDAC2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A3E4 | |
| Unconjugated | |
| 150 μL | |
| 3066 | |
| IgG2b |