Learn More
HDAC9 Monoclonal Antibody (OTI7G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805392S
Description
HDAC9 Monoclonal antibody specifically detects HDAC9 in Human samples. It is validated for Western BlotSpecifications
| HDAC9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15582 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7045 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7G9 | |
| Unconjugated | |
| TGFBI | |
| AV022454; D030072B18Rik; DKFZp779K1053; HD7; HD7b; HD9; HDAC; HDAC7; Hdac7b; HDAC9; hdac-9; HDAC9B; HDAC9FL; HDRP; histone deacetylase 4/5-related protein; histone deacetylase 7B; Histone deacetylase 9; histone deacetylase-related protein; KIAA0744; MEF-2 interacting transcription repressor (MITR) protein; MEF2-interacting transcription repressor MITR; Mitr; mKIAA0744; RGD1310748; RGD1563092; TWIST | |
| Human recombinant protein fragment corresponding to amino acids 405-683 of human TGFBI produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.