missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDGF Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148513
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
HDGF is a member of the hepatoma-derived growth factor family. This protein has mitogenic and DNA-binding activity and may play a role in cellular proliferation and differentiation.
Specifications
| HDGF | |
| Polyclonal | |
| Unconjugated | |
| HDGF | |
| AI118077; D3Ertd299e; DKFZp686J1764; FLJ96580; Hdgf; Hepatoma-derived growth factor; high mobility group protein 1-like 2; HMG1L2; HMG-1L2; Tdrm1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 3068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P51858 | |
| HDGF | |
| E. coli-derived recombinant human HDGF, Ser2-Lys240 (Pro101Lys). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction