missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HDGFRP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100100
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HRP2.
Hepatoma-derived growth factor-related protein 2 (HDGFRP2) is a widely expressed member of protein family that contains the HDGF related proteins 1-4 and lens epithelial derived growth factor (LEDGF). These family members show extensive homology at the amino terminus (termed the hath region). Little is known of the role of HDGFRP2, but like the related LEDGF, HDGFRP2 can bind HIV-1 integrase and stimulate HIV-1 integrase activity in vitro.
Specifications
| HDGFRP2 | |
| Polyclonal | |
| Unconjugated | |
| HDGFL2 | |
| HDGF2; HDGF-2; Hdgf3; HDGF-3; Hdgfl2; HDGF-related protein 2; HDGFRP2; Hepatoma-derived growth factor 2; Hepatoma-derived growth factor 3; hepatoma-derived growth factor, related protein 2; hepatoma-derived growth factor-related protein 2; HRP2; HRP-2; UNQ785/PRO1604 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15193, 171073, 84717 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UMU9, Q7Z4V5, Q925G1 | |
| HDGFL2 | |
| A synthesized peptide derived from human HDGFL2(Accession Q7Z4V5), corresponding to amino acid residues Y40-A90. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur