Learn More
Huenersdorf™ HDPE Industrial Bucket
Product Code. 11796715
Change view
Click to view available options
Capacity (Metric):
10.5 L
17 L
Color:
Natural
Description:
Round
Square
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Description unitSize
11796715 17 L Natural Round Each
11816124 10.5 L Natural Round Each
11863652 - Natural Square Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11796715 Supplier Huenersdorf™ Supplier No. 942100
  • ONLINE EXCLUSIVE PRICE
    51.39€
    10.53€ / Pack of 1
    Save 40.86€
    79% Off
In Stock
Add to basket
Add to basket
Use the Huenersdorf™ HDPE Industrial Bucket for all your laboratory needs. It is food-safe and has a spout for safe handling.
  • Round with spout
  • Natural
  • Acid-resistant polyethylene (HDPE)
  • Embossed scale and plastic handle Industrial bucket with spout for safe handling
Specifications
Product Type Industrial Bucket
Capacity (Metric) 17 L
Material HDPE
Color Natural
Description Round
Diameter (Metric) Outer 330 mm
Height (Metric) 315 mm
Includes Spout
Quantity 1 Ea.