Learn More
Ansell Kimtech™ M3 Sterile Face Masks
Product Code. 15691692
Change view
Click to view available options
Fastening Type:
Earloops
Ties
Includes:
7 in. Soft Ties
9 in. Soft Ties
Knitted Earloops
Unit Size:
Pack of 200
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fastening Type Includes Unit Size
15691692 Earloops Knitted Earloops Pack of 200
11715505 Ties 7 in. Soft Ties Pack of 200
11393892 Ties 9 in. Soft Ties Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15691692 Supplier Ansell Supplier No. 62470
  • ONLINE EXCLUSIVE PRICE
    472.00€
    316.47€ / Pack of 200
    Save 155.53€
    32% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Sterile masks feature non-irritating BiCoSof™ inner facing and bendable aluminum nosepiece. Available with earloops or soft ties.
  • Gamma-sterilized to 10-6
  • White with soft ties or earloops
  • Outer layer of apertured polyethylene film to provide protection from contamination
  • Polyester binding and polyester filter media provide bacteria filtration efficiency of 91.9% and particle filtration efficiency of 93.1%
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material SMS
Size Standard
Sterility Sterile
Style Pleated
Certifications/Compliance ISO 14001, ISO 9001; CE Simple; Industrial Use Only
Cleanroom Class Class 1 (ISO 3)
Disposable Single-use
Filter Efficiency 89.3% (PFE), 94.1% (BFE)
Includes Knitted Earloops
Latex Free Yes
Model 62470
Pore Size 0.1 μm (PFE), 3 μm (BFE)
Series Kimtech™ M3 Sterile