Learn More
Keystone Cleanroom Products Heat Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE
Product Code. 17777817 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
3 x 8 in.
6 x 3 in.
8 x 6 in.
10 x 8 in.
12 x 6 in.
15 x 8 in.
4 x 15 in.
6 x 25 in.
12 x 12 in.
17 x 12 in.
17 x 16 in.
18 x 12 in.
18 x 24 in.
20 x 10 in.
22 x 18 in.
24 x 12 in.
24 x 24 in.
30 x 10 in.
30 x 24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17777817 18 x 24 in. Pack of 250
17717807 6 x 3 in. Pack of 1000
17747807 8 x 6 in. Pack of 500
17757807 12 x 6 in. Pack of 500
17777807 10 x 8 in. Pack of 500
17787807 15 x 8 in. Pack of 500
17797807 20 x 10 in. Pack of 500
17707817 30 x 10 in. Pack of 500
17717817 12 x 12 in. Pack of 500
17727817 17 x 12 in. Pack of 500
17737817 18 x 12 in. Pack of 500
17747817 24 x 12 in. Pack of 500
17757817 17 x 16 in. Pack of 500
17767817 22 x 18 in. Pack of 500
17787817 24 x 24 in. Pack of 250
17797817 30 x 24 in. Pack of 250
17737807 4 x 15 in. Pack of 500
17767807 6 x 25 in. Pack of 500
17727807 3 x 8 in. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 17777817 Supplier Keystone Cleanroom Products Supplier No. BBMGTVKHS2418
Only null left
Add to basket
Add to basket
Heat Seal Tyvek™ Steam Sterilization Pouches provide a strong barrier from microbial contamination.
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing a strong barrier from microbial contamination
  • The clear film side offers high clarity along with strength and durability
  • Should be heat sealed closed using a validate-able impulse, or continuous band heat sealer
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Works with both Impulse and Continuous Band Sealers
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 250/Pk.
Dimensions (L x W) 18 x 24 in.
Length (English) 18 in.
Width (English) 24 in.
Product Type Autoclave Bag
Length (Metric) 45.7 cm
Width (Metric) 61 cm