Learn More
Carver™ Heated Manual Two Post Press
Product Code. 11900017
Change view
Click to view available options
Normal Force:
22 tons
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Normal Force Unit Size
11900017 22 tons Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Special packaging extra freight cost. Please contact us for quotation
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11900017 Supplier Carver™ Supplier No. 4389CE.4020000
  • ONLINE EXCLUSIVE PRICE
    51417.14€
    15813.43€ / Pack of 1
    Save 35603.71€
    69% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Special packaging extra freight cost. Please contact us for quotation
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
TheCarver™ model MH heated manual two post press is used for numerous applications for sample prep in the chemical, physical, biological and mechanical fields, wherever pressing is required.
  • 23 x 23 mm plates
  • <70 db(A)
  • Maximum pressure 669 bar/9709 psi
  • 165 mm Ram stroke
Specifications
Product Type Manual Press
For Use With (Application) R&D, Specialised Low Volume Development, Quality Testing
Weight (Metric) 425 kg
Normal Force 22 tons