Learn More
Horst™ Heating Cable, Quartz Yarn, High Temperature Resistance
Product Code. 10230542
Change view
Click to view available options
Length (Metric):
370 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10230542 370 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10230542 Supplier Horst™ Supplier No. 21002
  • ONLINE EXCLUSIVE PRICE
    217.35€
    144.43€ / Pack of 1
    Save 72.92€
    33% Off
View all promotions
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
High temperatures, small winding radii, high power rate.
  • Flexible.
  • High temperature resistance.
  • High power rates per meter.
  • Quartz yarn insulation.
  • Small winding radii.
Specifications
Description Bending radius: 10 mm, Thickness: 5 mm
Length (Metric) 370 m
Voltage 230 V Nominal
Material Quartz Yarn
Length (Metric) Power Cord 1.5 m
Wattage 2.1 W
Width (Metric) 3.5 to 4.5 mm
Temperature (Metric) 900°C (Nominal)