Learn More
Invitrogen™ HEATR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556559
Description
Immunogen sequence: KSEDTIDFLE FKYCVSLRTQ ICQALIHLLS LASASDLPCM KETLELSGNM VQSYILQFLK SGAEGDDTGA PHSPQERDQM VRMALKHMGS IQAPTGDTAR RAIMGFLEEI LAVCFDSSGS QGAL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
Amplification-dependent oncogene.
Specifications
| HEATR6 | |
| Polyclonal | |
| Unconjugated | |
| HEATR6 | |
| 2700008B19Rik; 2700008B19Rik-like protein; ABC1; amplified in breast cancer 1; amplified in breast cancer protein 1; BB135312; BB249351; HEAT repeat containing 6; HEAT repeat-containing protein 6; HEATR6; RGD1566204 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6AI08 | |
| HEATR6 | |
| Recombinant protein corresponding to Human HEATR6. Recombinant protein control fragment (Product #RP-93170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.